Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597663
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597663 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597663 Supplier Invitrogen™ Supplier No. PA597663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DLK1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DLK1
Gene Accession No. P80370
Gene Alias Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG
Gene Symbols DLK1
Host Species Rabbit
Immunogen Recombinant Human Protein delta homolog 1 protein (24-303AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114587, 8788
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.