Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569625 Supplier Invitrogen™ Supplier No. PA569625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 93%.

Regulates adipogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLK2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dlk2
Gene Accession No. Q8K1E3
Gene Alias AI413481; delta like non-canonical Notch ligand 2; delta-like 2 homolog; delta-like 2 homolog (Drosophila); DLK2; DLK-2; Egfl9; EGF-like protein 9; EGF-like-domain, multiple 9; Endothelial cell-specific protein S-1; epidermal growth factor-like protein 9; Protein delta homolog 2; UNQ2903/PRO28633
Gene Symbols Dlk2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse DLK2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106565
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.