Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL1 Antibody (019), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290620AF532
Description
DLL1 Monoclonal specifically detects DLL1 in Mouse samples. It is validated for ELISA.Specifications
DLL1 | |
Monoclonal | |
Alexa Fluor 532 | |
Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL1 (NP_031891.2; Met 1-Gln 516). | |
0.1 mL | |
Neuroscience, Stem Cell Signaling Pathway | |
28514 | |
Mouse | |
Purified |
ELISA | |
19 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human DLL1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction