Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL3 Antibody (7B7), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DLL3 |
---|---|
Clone | 7B7 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
DLL3 | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
Recombinant Human DLL3 protein [UniProt Q9NYJ7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7B7 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
Cell Biology, Cell Cycle and Replication, Neuroscience | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
10683 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title