Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523448
Description
Sequence homology: Cat (83%), Bovine (81%), Mouse, Rat (72%).
Delta-like protein 3 (DLL3) is a ligand for the Notch signaling pathway. It inhibits primary neurogenesis. DLL3 plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. Defects in DLL3 are the cause of Spondylocostal dysostosis autosomal recessive type 1 (SCDO1). Mutations in DLL3 gene cause truncal shortening relative to their limbs, which leads to abdominal protrusion, abnormal spinal curvature and sometimes a plagiocephaly-torticollis sequence. It may be required to divert neurons along a specific differentiation pathway.
Specifications
| DLL3 | |
| Polyclonal | |
| Unconjugated | |
| Dll3 | |
| delta like canonical Notch ligand 3; Delta3; delta-like 3; delta-like 3 (Drosophila); delta-like protein 3; Dll3; Drosophila Delta homolog 3; M-Delta-3; pu; pudgy; SCDO1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10683, 13389 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O88516, Q9NYJ7 | |
| Dll3 | |
| A portion of amino acid 100 to 150 of human DLL3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction