Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL4 Antibody (004), DyLight 594 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290662DL594
Description
DLL4 Monoclonal antibody specifically detects DLL4 in Mouse samples. It is validated for ELISA, ELISASpecifications
| DLL4 | |
| Monoclonal | |
| DyLight 594 | |
| delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL4 (NP_062327.2; Met1-Pro525). | |
| 0.1 mL | |
| Neuroscience | |
| 54567 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 004 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction