Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551794 Supplier Invitrogen™ Supplier No. PA551794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNTCAMLTTF NEIDMSNIQE MRARHKEAFL KKHNLKLGFM SAFVKASAFA LQEQPVVNAV IDDTTKEVVY RDYIDISVAV ATPRGLVVPV IRNVEAMNFA DIERTITELG EKARKNELAI EDMDGGTFTI SNGGVFGSLF GTPII Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes a mitochondrial protein that belongs to the 2-oxoacid dehydrogenase family. This protein is one of the three components (the E2 component) of the 2-oxoglutarate dehydrogenase complex that catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dlst
Gene Accession No. P36957, Q01205, Q9D2G2
Gene Alias 1600017E01Rik; 2-oxoglutarate dehydrogenase complex component E2; 4632413C10Rik; 4930529O08Rik; dihydrolipoamide S-succinyltransferase; dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex); dihydrolipoamide succinyltransferase component of 2-oxoglutarate dehydrogenase complex; Dihydrolipoyllysine-residue succinyltransferase; dihydrolipoyllysine-residue succinyltransferase component of 2-oxoglutarate dehydrogenase complex, mitochondrial; DLST; DLTS; E2; E2K; OGDC-E2
Gene Symbols Dlst
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DLST. Recombinant protein control fragment (Product #RP-88908).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1743, 299201, 78920
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.