Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dlx1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216178
Description
Dlx1 Polyclonal specifically detects Dlx1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Dlx1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
distal-less homeo box 1, distal-less homeobox 1, homeobox protein DLX-1 | |
Rabbit | |
27 kDa | |
0.1 mL | |
Neuroscience | |
1745 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:100-1:1000 | |
P56177 | |
DLX1 | |
Full length human Dlx1 Recombinant protein. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (98%), Chicken (89%), Rat (98%), Bovine (98%), Xenopus tropicalis (85%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction