Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583799 Supplier Invitrogen™ Supplier No. PA583799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTMTTMPESL NSPVSGKAVF MEFGPPNQQM SPSPMSHGHY SMHCLHSAGH SQPDGAYSSA SSFSRPLGYP YVNSVS Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-beta superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DLX1
Gene Accession No. P56177
Gene Alias DII B; distal-less homeo box 1; distal-less homeobox 1; Dlx; Dlx1; Dlx-1; homeobox Dlx1; homeobox protein DLX-1
Gene Symbols DLX1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human DLX1 (Product #RP-96565)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1745
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.