Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540408 Supplier Invitrogen™ Supplier No. PA540408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MTGVFDSLVA DMHSTQIAAS STYHQHQQPP SGGGAGPGGN SSSSSSLHKP Sequence homology: Cow: 87%; Dog: 92%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 92%.

Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DLX2
Gene Accession No. P40764, Q07687
Gene Alias AW121999; DII A; distal-less homeo box 2; distal-less homeobox 2; DLX2; Dlx-2; Dlx5; DLX-5; homeobox Dlx5; homeobox protein DLX-2; Homeobox protein TES-1; TES1; Tes-1
Gene Symbols DLX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DLX2 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13392, 1746
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.