Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111443
Description
Immunogen sequence: RRSKFKKMWK SGEIPSEQHP GASASPPCAS PPVSAPASWD FGVPQRM.
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2.
Specifications
| DLX2 | |
| Polyclonal | |
| Unconjugated | |
| DLX2 | |
| AW121999; DII A; distal-less homeo box 2; distal-less homeobox 2; DLX2; Dlx-2; Dlx5; DLX-5; homeobox Dlx5; homeobox protein DLX-2; Homeobox protein TES-1; TES1; Tes-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1746 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q07687 | |
| DLX2 | |
| Recombinant Protein corresonding to Human DLX2. Recombinant protein control fragment (Product #RP-106743). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction