Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLX3 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297552AF700
Description
DLX3 Polyclonal specifically detects DLX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DLX3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AI4, distal-less homeo box 3, distal-less homeobox 3, homeobox protein DLX-3, TDO | |
Produced in rabbits immunized with E. coli-derived Human DLX3 fragment. | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Stem Cell Markers | |
1747 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction