Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 132613AP150UL
Description
The distal-less homeobox 3 (DLX3) protein is a putative transcriptional activator that has been implicated during development and differentiation of epithelial tissues. This is a homeodomain transcription factor in vertebrates, related to Distal-less in Drosophila, which is expressed in differentiating epidermal cells, in neural crest, hair follicles, dental epithelium, mesenchyme, the optic and olfactory placodes, limb bud, placenta, and in the cement gland, which is located in the extreme anterior neural plate in Xenopus embryos.Specifications
| DLX3 | |
| Polyclonal | |
| Unconjugated | |
| DLX3 | |
| AI4, distal less homeobox 3, DLX3, Homeobox protein DLX 3, TDO | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13393, 1747, 287638 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60479, Q64205 | |
| Dlx3 | |
| DLX3 Fusion Protein Ag3865 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction