Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLX4 Monoclonal Antibody (OTI7C8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527027
Description
DLX4 Monoclonal Antibody for Western Blot, IHC (P)
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant has not been described. Studies of the two splice variants revealed that one encoded isoform functions as a repressor of the beta-globin gene while the other isoform lacks that function.
Specifications
DLX4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92988 | |
DLX4 | |
Full length human recombinant protein of DLX4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7C8 | |
Unconjugated | |
DLX4 | |
beta protein 1; BP1; DII D; distal-less homeo box 7; distal-less homeo box 9; distal-less homeobox 4; DLX4; Dlx-4; Dlx7; DLX8; DLX9; homeobox protein DLX-4; Homeobox protein DLX-7; homeobox protein DLX-8; OFC15 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1748 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction