Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563798 Supplier Invitrogen™ Supplier No. PA563798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLSYPYTEPA NPGDSYLSCQ QPAALSQPLC GPAEHPQELE ADSEKPRLSP EPS Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 69%.

Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant has not been described. Studies of the two splice variants revealed that one encoded isoform functions as a repressor of the beta-globin gene while the other isoform lacks that function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DLX4
Gene Accession No. Q92988
Gene Alias beta protein 1; BP1; DII D; distal-less homeo box 7; distal-less homeo box 9; distal-less homeobox 4; DLX4; Dlx-4; Dlx7; DLX8; DLX9; homeobox protein DLX-4; Homeobox protein DLX-7; homeobox protein DLX-8; OFC15
Gene Symbols DLX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DLX4. Recombinant protein control fragment (Product #RP-101173).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.