Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101134 Supplier Invitrogen™ Supplier No. PA5101134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DLX5.

This gene encodes a member of a homeobox transcription factor gene family similar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DLX5
Gene Accession No. P50575, P56178, P70396
Gene Alias AI385752; distal-less homeo box 5; distal-less homeobox 5; distal-less homeobox protein 5; DLX-3; DLX5; Homeobox protein DLX-3; homeobox protein DLX-5; RDLX; SHFM1D; split hand/foot malformation type 1 with sensorineural hearing loss; Unknown (protein for MGC:137934)
Gene Symbols DLX5
Host Species Rabbit
Immunogen A synthesized peptide derived from human DLX5(Accession P56178), corresponding to amino acid residues G106-F156.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13395, 1749, 25431
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.