Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540409
Description
Peptide sequence: NPYQYQYHGV NGSAGSYPAK AYADYSYASS YHQYGGAYNR VPSATNQPEK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.
This gene encodes a member of a homeobox transcription factor gene family similar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation.
Specifications
DLX5 | |
Polyclonal | |
Unconjugated | |
DLX5 | |
AI385752; distal-less homeo box 5; distal-less homeobox 5; distal-less homeobox protein 5; DLX-3; DLX5; Homeobox protein DLX-3; homeobox protein DLX-5; RDLX; SHFM1D; split hand/foot malformation type 1 with sensorineural hearing loss; Unknown (protein for MGC:137934) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13395, 1749 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P56178, P70396 | |
DLX5 | |
Synthetic peptide directed towards the N-terminal of human DLX5 (aa 71-120). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction