Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLX5 Rabbit anti-Human, Clone: 2F9C6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31660520UL
This item is not returnable.
View return policy
Description
DLX5 Monoclonal antibody specifically detects DLX5 in Human samples. It is validated for Western BlotSpecifications
DLX5 | |
Monoclonal | |
Unconjugated | |
distal-less homeo box 5, distal-less homeobox 5, homeobox protein DLX-5 | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of human DLX5 (P56178). | |
20 μg | |
Neuroscience | |
1749 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
2F9C6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction