Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dlx6 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297544JF646
Description
Dlx6 Polyclonal specifically detects Dlx6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Dlx6 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
distal-less homeo box 6, distal-less homeobox 6, homeobox protein DLX-6, MGC125282, MGC125283, MGC125284, MGC125285 | |
Produced in rabbits immunized with E. coli-derived Human Dlx6 fragment. | |
0.1 mL | |
Neuroscience | |
1750 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction