Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dlx6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $646.00
Specifications
Antigen | Dlx6 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Dlx6 Polyclonal specifically detects Dlx6 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Dlx6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay | |
Unconjugated | |
RUO | |
Human | |
distal-less homeo box 6, distal-less homeobox 6, homeobox protein DLX-6, MGC125282, MGC125283, MGC125284, MGC125285 | |
DLX6 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
1750 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ALPERAELAASLGLTQTQVKIWFQNKRSKFKKLLKQGSNPHESDPLQGSAALSPRSPALPPVWDVSASAKGVSMPPNSYMPGYSHWYSSPHQDTMQRPQMM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title