Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566396
Description
Immunogen sequence: GNSYNHRSLAA YPYMSHSQHS PYLQSYHNSS AAAQTRGDDT DQQKTTVIEN GEIRF Highest antigen sequence identity to the following orthologs - mouse 100%, rat 75%.
The DLX6 gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. This family is comprised of at least 6 different members that encode proteins with roles in forebrain and craniofacial development. This gene is in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7.
Specifications
DLX6 | |
Polyclonal | |
Unconjugated | |
DLX6 | |
distal-less homeo box 6; distal-less homeobox 6; distal-less homeobox 6-like; DLX6; Homeobox protein DLX-6; RGD1561539 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1750 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P56179 | |
DLX6 | |
Recombinant Human DLX6. Recombinant protein control fragment (Product #RP-107070). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction