Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596753 Supplier Invitrogen™ Supplier No. PA596753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSHSQHSPYL QSYHNSSAAA QTRGDDTDQQ KTTVIENGEI RFNGKGKKIR KPRTIYSSLQ LQALNHRFQQ TQYLALPERA ELAASLGLTQ TQVKIWFQNK RSKFKKLLKQ GSNPHESDPL QGSAALSPRS PALPPVWDVS ASAKGVSMPP NSYMPGYSHW YSSPHQDTMQ RPQMM; Positive Samples: SH-SY5Y, Jurkat, HeLa, 293T; Cellular Location: Nucleus.

The DLX6 gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. This family is comprised of at least 6 different members that encode proteins with roles in forebrain and craniofacial development. This gene is in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DLX6
Gene Accession No. P56179
Gene Alias distal-less homeo box 6; distal-less homeobox 6; distal-less homeobox 6-like; DLX6; Homeobox protein DLX-6; RGD1561539
Gene Symbols DLX6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 119-293 of human DLX6 (NP_0052133).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1750
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.