Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572622
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572622 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572622 Supplier Invitrogen™ Supplier No. PA572622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DLX6A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX6A
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene dlx6a
Gene Accession No. Q98877
Gene Alias distal-less homeobox 6a; distal-less homeobox gene 6a; distal-less homeobox protein 6a; Dlx6; DLX-6; dlx6a; Homeobox protein Dlx6a
Gene Symbols dlx6a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 167-194 amino acids from the C-terminal region of DANRE dlx6a.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30586
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.