Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMAP1 Antibody, HRP, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100256H
Description
DMAP1 Polyclonal antibody specifically detects DMAP1 in Human samples. It is validated for Western BlotSpecifications
DMAP1 | |
Polyclonal | |
Western Blot | |
DNA methyltransferase 1 associated protein 1, DNA methyltransferase 1-associated protein 1, DNMAP1DNMT1-associated protein 1, DNMT1 associated protein 1, DNMTAP1, EAF2, FLJ11543, KIAA1425DKFZp686L09142, MEAF2, SWC4 | |
The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human DNA Methyltransferase 1 Associated Protein 1 using the numbering given in entry NP_061973.1 (GeneID 55929). | |
0.1 mL | |
Apoptosis, Chromatin Research, DNA Repair, MAP Kinase Signaling | |
55929 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction