Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMAP1 Goat anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557127
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-155-7127 100 μL
1 options

Catalog No. 50-155-7127

Supplier: Bethyl Laboratories, Inc A300217A

Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

DNA methyltransferase 1-associated protein 1 (DMAP1) is involved in the repression or activation of transcription. The encoded protein can independently repress transcription and is targeted to replication foci throughout S phase by interacting directly with the N-terminus of DNA methyltransferase 1. During late S phase, histone deacetylase 2 is added to this complex, providing a means to deacetylate histones in transcriptionally inactive heterochromatin following replication. The protein also specifically potentiates DAXX-mediated repression of glucocorticoid receptor-dependent transcription. The encoded protein is also a component of the nucleosome acetyltransferase of H4 complex and interacts with the transcriptional corepressor tumor susceptibility gene 101 and the pro-apoptotic death-associated protein 6, among others.DMAP1 is found in a wide variety of living organisms from yeasts to humans. The human DMAP1 gene is localizedon chromosome 1p34.

Specifications

Antigen DMAP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DMAP1
Gene Accession No. Q9NPF5
Gene Alias RP5-891H21.2, DKFZp686L09142, DNMAP1, DNMTAP1, EAF2, FLJ11543, KIAA1425, MEAF2, SWC4
Gene Symbols DMAP1
Host Species Goat
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55929
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.