Learn More
DMAP1 Goat anti-Human, Polyclonal, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300218A
Description
The recommended shelf life for this product is 1 year from date of receipt.
DNA methyltransferase 1-associated protein 1 (DMAP1) is involved in the repression or activation of transcription. The encoded protein can independently repress transcription and is targeted to replication foci throughout S phase by interacting directly with the N-terminus of DNA methyltransferase 1. During late S phase, histone deacetylase 2 is added to this complex, providing a means to deacetylate histones in transcriptionally inactive heterochromatin following replication. The protein also specifically potentiates DAXX-mediated repression of glucocorticoid receptor-dependent transcription. The encoded protein is also a component of the nucleosome acetyltransferase of H4 complex and interacts with the transcriptional corepressor tumor susceptibility gene 101 and the pro-apoptotic death-associated protein 6, among others.DMAP1 is found in a wide variety of living organisms from yeasts to humans. The human DMAP1 gene is localizedon chromosome 1p34.Specifications
| DMAP1 | |
| Polyclonal | |
| Unconjugated | |
| DMAP1 | |
| RP5-891H21.2, DKFZp686L09142, DNMAP1, DNMTAP1, EAF2, FLJ11543, KIAA1425, MEAF2, SWC4 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 55929 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9NPF5 | |
| DMAP1 | |
| Between 400 and 450 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.