Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMBT1/GP340 Antibody (548031), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5915K
Description
DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
DMBT1/GP340 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
548031 | |
Flow Cytometry, CyTOF-ready | |
deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein | |
E. Coli-derived recombinant mouse DMBT1, Trp215-Gly420, Accession # Q60997 | |
0.1 mL | |
Breast Cancer | |
1755 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction