Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMBX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568546
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Rabbit: 79%; Rat: 93%.
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
DMBX1 | |
Polyclonal | |
Unconjugated | |
DMBX1 | |
Atx; Cdmx; diencephalon/mesencephalon homeobox 1; diencephalon/mesencephalon homeobox protein 1; Diencephalon/mesencephalon-expressed brain homeobox gene 1 protein; dmbx1; homeobox gene Atx; homeoprotein MBX; MBX; Orthodenticle homolog 3; OTX; OTX3; Paired-like homeobox protein DMBX1; paired-type homeobox Atx; PaxB | |
Rabbit | |
Affinity chromatography | |
RUO | |
127343 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NFW5 | |
DMBX1 | |
synthetic peptide directed towards the middle region of human DMBX1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction