Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMBX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569044
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
DMBX1 | |
Polyclonal | |
Unconjugated | |
DMBX1 | |
Atx; Cdmx; diencephalon/mesencephalon homeobox 1; diencephalon/mesencephalon homeobox protein 1; Diencephalon/mesencephalon-expressed brain homeobox gene 1 protein; dmbx1; homeobox gene Atx; homeoprotein MBX; MBX; Orthodenticle homolog 3; OTX; OTX3; Paired-like homeobox protein DMBX1; paired-type homeobox Atx; PaxB | |
Rabbit | |
Affinity chromatography | |
RUO | |
140477 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q91ZK4 | |
DMBX1 | |
synthetic peptide directed towards the C-terminal region of mouse DMBX1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction