Learn More
Invitrogen™ DMBX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555425
Description
Immunogen sequence: SLTITPVAPG GGLLGPSHSY SSSPLSLFRL QEQFRQHMAA TNNLVHYSSF EVGGPAPAAA AAAAAVPYLG VNMAPLGSLH CQSYYQSLSA AAAAHQGVWG SPLLPAPPAG LAPASATLNS KTTSIENLRL RAKQHAASLG LDTLP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
DMBX1 | |
Polyclonal | |
Unconjugated | |
DMBX1 | |
Atx; Cdmx; diencephalon/mesencephalon homeobox 1; diencephalon/mesencephalon homeobox protein 1; Diencephalon/mesencephalon-expressed brain homeobox gene 1 protein; dmbx1; homeobox gene Atx; homeoprotein MBX; MBX; Orthodenticle homolog 3; OTX; OTX3; Paired-like homeobox protein DMBX1; paired-type homeobox Atx; PaxB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFW5 | |
DMBX1 | |
Recombinant protein corresponding to Human DMBX1. Recombinant protein control fragment (Product #RP-89442). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.