Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521283

Catalog No. PIPA521283


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (98%), Zebrafish (96%), Japanese Medaka (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DMC1
Polyclonal
Unconjugated
DMC1
DCM1; disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; dJ199H16.1; DMC 1; DMC1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; DMC1H; DNA meiotic recombinase 1; HsLim15; LIM15; Mei11; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4; sgdp
Rabbit
Antigen Affinity Chromatography
RUO
11144
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q14565
DMC1
Recombinant fragment corresponding to a region within amino acids 168 and 340 of DMC1 (Uniprot ID#Q14565).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.