Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521472 Supplier Invitrogen™ Supplier No. PA521472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa, Molt-4, Raji, mouse testis. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (83%), Japanese Medaka (84%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DMC1
Gene Accession No. Q14565, Q61880
Gene Alias DCM1; disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; dJ199H16.1; DMC 1; DMC1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; DMC1H; DNA meiotic recombinase 1; HsLim15; LIM15; Mei11; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4; sgdp
Gene Symbols DMC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DMC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11144, 13404
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.