Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581926
Description
Immunogen sequence: AYTSEHQMEL LDYVAAKFHE EAGIFKLLII DSIMALFRVD FSGRGELAER QQKLAQMLSR LQKISEEYNV AVFVTNQMTA DPGATMTFQA DPKKPIGGHI LAHASTTRIS LRKGRGELRI AKIYDSPEMP ENEATFAITA.
DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
Specifications
DMC1 | |
Polyclonal | |
Unconjugated | |
DMC1 | |
DCM1; disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; dJ199H16.1; DMC 1; DMC1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; DMC1H; DNA meiotic recombinase 1; HsLim15; LIM15; Mei11; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4; sgdp | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11144 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14565 | |
DMC1 | |
Recombinant protein corresponding to Human DMC1. Recombinant protein control fragment (Product #RP-91372). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction