Learn More
Thermo Scientific™ DMEM for SILAC, 6 x 500 mL PROMO
Description
Features of DMEM for SILAC:
- Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
- High quality—medium is sterile, endotoxin-free, and cell culture-compatible
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.
This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.
Related products
- NeuCode™ Lysine -080, 50 mg (A33613)
- Fetal Bovine Serum, dialyzed, US origin (26400036)
- RPMI 1640 Media for SILAC (88365)
Specifications
Specifications
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | Media |
| Content And Storage | Store at 2°C to 8°C. |
| Label or Dye | Mass Isotope of Amino Acid |
| Shipping Condition | Room Temperature |
| Starting Material | Cell Lysate |
| Final Product Type | Liquid Medium |
| Quantity | 6 x 500 mL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.