Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMF, Dimethylformamide, Sequencing, Fisher BioReagents

Catalog No. p-25570 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle

Catalog No. BP1160-4

Only null left
Add to Cart
  • DMF for sequencing and peptide synthesis
  • Useful for protein chemistry due to low amine content
  • DMF is a commonly used solvent in solid phase peptide synthesis
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 6 to 8
Melting Point -61°C
Absorbance Pass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm
Color Colorless
Decomposition Information >350°C
Density 0.945g/cm³
Boiling Point 153°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.0005mEq/g max.
Linear Formula HCONCH3CH3
Titratable Base 0.003mEq/g max.
Vapor Pressure 4.9mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
UN Number 2265
Water 0.03% max.
Merck Index 15, 3267
Formula Weight 73.09
Percent Purity ≥99.5%
Grade Sequencing
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.