Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557660 Supplier Invitrogen™ Supplier No. PA557660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLATDDVDPE GNESIWYNGK VVGNTTSGSY SYSIQKSLAF AYVPVQLSEV GQQVEVELLG KNYPAVIIQE PLVLTEPTRN RLQK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMGDH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMGDH
Gene Accession No. Q9UI17
Gene Alias 1200014D15Rik; AI787269; dimethylglycine dehydrogenase; dimethylglycine dehydrogenase precursor; dimethylglycine dehydrogenase, mitochondrial; Dmgdh; DMGDHD; EC 1.5.99.2; M2GD; ME2GLYDH
Gene Symbols DMGDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMGDH. Recombinant protein control fragment (Product #RP-96850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.