Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMKN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545803 Supplier Invitrogen™ Supplier No. PA545803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QPGAGWQEVA AVTSKNYNYN QHAYPTAYGG KYSVKTPAKG GVSPSSSASR Sequence homology: Cow: 86%; Dog: 86%; Human: 100%; Rabbit: 86%.

The function of this protein is still unknown. The human ZD52F10 gene is up-regulated in colon cancer metastasis. It encode the protein (dermokine), which is a new stratified epithelium-secreted protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMKN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dmkn
Gene Accession No. Q6E0U4
Gene Alias 1110014F24Rik; AW561900; C130074A08; cI-36; Dermokine; Dmkn; Epidermis-specific secreted protein SK30/SK89; RGD1561521; SK30; SK89; UNQ729; UNQ729/PRO1411; ZD52F10
Gene Symbols Dmkn
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DMKN.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93099
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.