Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMKN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5100241

Description
Antibody detects endogenous levels of total DMKN.
The function of this protein is still unknown. The human ZD52F10 gene is up-regulated in colon cancer metastasis. It encode the protein (dermokine), which is a new stratified epithelium-secreted protein.
Specifications
DMKN | |
Polyclonal | |
Unconjugated | |
Dmkn | |
1110014F24Rik; AW561900; C130074A08; cI-36; Dermokine; Dmkn; Epidermis-specific secreted protein SK30/SK89; RGD1561521; SK30; SK89; UNQ729; UNQ729/PRO1411; ZD52F10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
361548, 73712, 93099 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6E0U4, Q6P253 | |
Dmkn | |
A synthesized peptide derived from human DMKN(Accession Q6E0U4), corresponding to amino acid residues T403-K453. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction