Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMP-1 Antibody (001), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290020G
Description
DMP-1 Monoclonal specifically detects DMP-1 in Human samples. It is validated for ELISA.Specifications
DMP-1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1758 | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DMP-1 (Q13316-1; Met1-Tyr513). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human CD44 Human CFH | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction