Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103323 Supplier Thermo Scientific Supplier No. PA5103323
Add to Cart
Add to Cart

DMP1 Polyclonal Antibody

DMP1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DMP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dmp1
Gene Accession No. O55188, Q13316
Gene Alias AG1; ARHP; ARHR; AV020965; dentin matrix acidic phosphoprotein 1; dentin matrix protein 1; DENTMAT; Dmp; DMP 1; Dmp1; DMP-1; MGC130441; PP; serine rich acidic phosphoprotein
Gene Symbols Dmp1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DMP1(Accession Q13316), corresponding to amino acid residues D478-Y513.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13406, 1758
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.