Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542121 Supplier Invitrogen™ Supplier No. PA542121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TRLGRGGAGD FRTHPFFFGL DWDGLRDSVP PFTPDFEGAT DTCNFDLVED Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

DMPK is a serine/threonine kinase expressed in various muscle and nerve tissues. Mutations in the untranslated region of the DMPK gene are a characteristic of myotonic dystrophy. The protein encoded by this gene is a serine-threonine kinase that is closely related to other kinases that interact with members of the Rho family of small GTPases. Substrates for this enzyme include myogenin, the beta-subunit of the L-type calcium channels, and phospholemman. The 3' untranslated region of this gene contains 5-37 copies of a CTG trinucleotide repeat. Expansion of this unstable motif to 50-5,000 copies causes myotonic dystrophy type I, which increases in severity with increasing repeat element copy number. Repeat expansion is associated with condensation of local chromatin structure that disrupts the expression of genes in this region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMPK
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DMPK
Gene Accession No. Q09013
Gene Alias DM; DM protein kinase; DM1; DM1 protein kinase; Dm15; DM1PK; DMK; DM-kinase; Dmpk; dystrophia myotonica kinase, B15; dystrophia myotonica protein kinase; dystrophia myotonica-protein kinase; MDPK; MT-PK; myotonic dystrophy associated protein kinase; myotonic dystrophy protein kinase; myotonin protein kinase A; myotonin-protein kinase; thymopoietin homolog
Gene Symbols DMPK
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DMPK.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1760
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.