Learn More
DMPO Nitrone Adduct Antibody (N1664A), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259306C
Catalog No. NBP259306C
Only null left
Add to Cart
Description
Description
DMPO Nitrone Adduct Monoclonal specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
DMPO Nitrone Adduct
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein G purified
Primary
Mouse, Non-species specific
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
N1664A
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide
5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin
100 μL
Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.