Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMPO Nitrone Adduct Antibody (N1664A), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259306IR
Description
DMPO Nitrone Adduct Monoclonal specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DMPO Nitrone Adduct | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Mouse, Non-species specific | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
N1664A | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide | |
5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin | |
100 μL | |
Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction