Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT1 Antibody (OTI1G10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246464
Description
Ensure accurate, reproducible results in Western Blot
DMRT1 Monoclonal specifically detects DMRT1 in Human samples. It is validated for Western Blot.Specifications
DMRT1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1 | |
Mouse | |
39.3 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI1G10 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
DMRT1 | |
Human recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1(NP_068770) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
1761 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction