Learn More
DMRT1 Mouse anti-Human, Clone: OTI1G10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807517

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.Specifications
DMRT1 | |
Monoclonal | |
Unconjugated | |
DMRT1 | |
CT154, DMT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1761 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1G10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5R6 | |
DMRT1 | |
Human recombit protein fragment corresponding to amino acids 1-210 of human DMRT1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.