Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597042 Supplier Invitrogen™ Supplier No. PA597042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHPIPLPSAA ELLVKRENNG SNPCLMTECS GTSQPPPASV PTTAASEGRM VIQDIPAVTS RGHVENTPDL VSDSTYYSSF YQPSLFPYYN NLYNCPQYSM ALAADSASGE VGNPLGGSPV KNSLRGLPGP YVPGQTGNQW QMKNMENRHA MSSQYRMHSY YPPPSYLGQS VPQFFTFEDA PSYPEARASV FSPPSSQDSG LVSLSSSSPI SNKSTKAVLE CEPASEPSSF TVTPVIEEDE; Positive Samples: HL-60; Cellular Location: Nucleus.

This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DMRT1
Gene Accession No. Q9Y5R6
Gene Alias CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119)
Gene Symbols DMRT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 134-373 of human DMRT1 (NP_0687702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1761
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.