Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31552
Description
Sequence: LAADSASGEVGNPLGGSPVKNSLRGLPGPYVPGQTGNQWQMKNMENRHAMSSQYRMHSYYPPPSYLGQSVPQFFTFEDAPSYPEARASVFSPPSSQDSGLVSLSSSSPISNKSTKAVLECEPASEPSSFTVTPVISpecifications
DMRT1 | |
Polyclonal | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:2500) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
doublesex and mab-3 related transcription factor 1 | |
DMRT1 | |
Recombinant protein corresponding to human DMRT1. | |
RUO | |
1761 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
DMT1 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction