Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT2 Antibody (PCRP-DMRT2-1B11), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314270IR
This item is not returnable.
View return policy
Description
DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
DMRT2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
10655 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
PCRP-DMRT2-1B11 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein | |
Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction