Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT2 Antibody (PCRP-DMRT2-1B11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314270F
This item is not returnable.
View return policy
Description
DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
DMRT2 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
10655 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, Immunofluorescence | |
PCRP-DMRT2-1B11 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein | |
Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction