Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRT2 Mouse anti-Human, Clone: PCRP-DMRT2-1B11, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | DMRT2 |
---|---|
Clone | PCRP-DMRT2-1B11 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array |
Applications | Western Blot, Flow Cytometry, Peptide Array |
Description
DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein ArraySpecifications
DMRT2 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Q9Y5R5 | |
10655 | |
IgG2b | |
Protein A or G purified |
PCRP-DMRT2-1B11 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein | |
Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title